Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023015.hg.1 |
NR_001562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023015.hg.1 |
TC04001655.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0400183498.hg.1 |
NR_001562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183498.hg.1 |
TC0400012165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400183498.hg.1 |
NR_001562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183499.hg.1 |
ENST00000505539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183499.hg.1 |
NR_001562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183499.hg.1 |
TC0400012165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400183499.hg.1 |
ENST00000505539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183499.hg.1 |
NR_001562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183500.hg.1 |
NR_001562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183500.hg.1 |
TC0400012165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400183500.hg.1 |
NR_001562.2 |
Refseq |