Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01674422_g1
          See other DEFA8P GE Assays ›
          Gene Symbol:
          DEFA8P
          Gene Name:
          defensin alpha 8, pseudogene
          Gene Aliases:
          DEFAP1
          Chromosome Location:
          Chr.8: 6950726 - 6951599 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01674422_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DEFA8P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_073407.1 1 - 2 169 96
          GenBank mRNA AY746433.1 - 1 - 2 169 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream defensin alpha 4 DEFA4 DEF4,
          HNP-4,
          HP-4,
          HP4
          -12.4kb Sense Chr. 8 - 6935820 - 6938338 on NCBI Build GRCh38
          Downstream defensin alpha 9,
          pseudogene
          DEFA9P DEFAP2 7.7kb Sense Chr. 8 - 6959289 - 6960161 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800009434.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214744 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214757 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941260 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214733 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16059278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16059275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941221 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941266 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214750 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941290 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214716 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214742 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941289 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214759 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05941238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214680 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3743432 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16546 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3743432 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01857526 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800009434.hg.1 ENST00000457582.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009434.hg.1 ENST00000457582.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009434.hg.1 NR_073407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009434.hg.1 NR_073407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164578.hg.1 ENST00000457582.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164578.hg.1 NR_073407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164578.hg.1 TC0800009434.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800164578.hg.1 NR_073407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164578.hg.1 ENST00000457582.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164577.hg.1 ENST00000457582.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164577.hg.1 NR_073407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164577.hg.1 TC0800009434.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800164577.hg.1 ENST00000457582.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164577.hg.1 NR_073407.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009434.hg.1 ENST00000457582.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009434.hg.1 ENST00000457582.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009434.hg.1 NR_073407.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009434.hg.1 NR_073407.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-55c5c7589b-pgs97:80/100.66.130.250:80.
          git-commit: fefcbad4eb9baa7f43a2c44c49530f643471c973
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0