Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305395.1 | NP_001292324.1 | 2 - 3 | 171 | 231 | |
NM_001305396.1 | NP_001292325.1 | 2 - 3 | 160 | 231 | ||
NM_001305397.1 | NP_001292326.1 | 2 - 3 | 171 | 204 | ||
XM_017009029.1 | XP_016864518.1 | 2 - 3 | 1358 | 231 | ||
XM_017009030.1 | XP_016864519.1 | 2 - 3 | 1357 | 204 | ||
GenBank mRNA | BU561730.1 | - | 2 - 3 | 107 | 231 | |
HY000873.1 | - | 1 - 2 | 83 | 231 | ||
HY001392.1 | - | 1 - 2 | 94 | 204 |
Set Membership: |
![]() ![]() ![]() ![]() |