Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000412001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
ENST00000412001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005473.hg.1 |
ENST00000282259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005473.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005473.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005473.hg.1 |
TC05000389.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500153941.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153941.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153941.hg.1 |
TC0500007881.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500153941.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153941.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000396137.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000412001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
ENST00000412001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007881.hg.1 |
NM_152405.4 |
Refseq |