Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146032.1 | NP_001139504.1 | 15 - 16 | 1272 | 138 | |
NM_138782.2 | NP_620137.2 | 16 - 17 | 1371 | 138 | ||
XM_017009016.1 | XP_016864505.1 | 15 - 16 | 1250 | 138 | ||
XM_017009017.1 | XP_016864506.1 | 16 - 17 | 1365 | 138 | ||
XM_017009018.1 | XP_016864507.1 | 16 - 17 | 1365 | 138 | ||
XM_017009019.1 | XP_016864508.1 | 16 - 17 | 1365 | 138 | ||
GenBank mRNA | AK291782.1 | - | 16 - 17 | 1301 | 138 | |
AK295141.1 | - | 15 - 16 | 1182 | 138 | ||
AL831971.1 | - | 16 - 17 | 1280 | 138 | ||
AL833313.1 | - | 13 - 14 | 975 | 138 | ||
BC137070.1 | - | 16 - 17 | 1371 | 9021067 | 138 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |