Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01560094_g1
          See other TNFRSF10A GE Assays ›
          Gene Symbol:
          TNFRSF10A
          Gene Name:
          TNF receptor superfamily member 10a
          Gene Aliases:
          APO2, CD261, DR4, TRAILR-1, TRAILR1
          Chromosome Location:
          Chr.8: 23191457 - 23225167 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01560094_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNFRSF10A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003844.3 NP_003835.3 7 - 8 933 67
          GenBank mRNA BC012866.1 - 7 - 8 892 3857315 67
          BT006906.1 - 7 - 8 827 67
          FJ795027.1 - 7 - 8 826 67
          JF729355.1 - 6 - 7 353 67
          U90875.1 - 7 - 8 827 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800009875.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08001052.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNFRSF10A

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          immune response
          signal transduction
          cell surface receptor signaling pathway
          activation of NF-kappaB-inducing kinase activity
          multicellular organism development
          extrinsic apoptotic signaling pathway via death domain receptors
          response to lipopolysaccharide
          tumor necrosis factor-mediated signaling pathway
          TRAIL-activated apoptotic signaling pathway
          regulation of cell population proliferation
          regulation of apoptotic process
          cellular response to mechanical stimulus
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          protease binding
          tumor necrosis factor-activated receptor activity
          death receptor activity
          protein binding
          transcription factor binding
          signaling receptor activity
          TRAIL binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000519862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000519862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000519862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013385.hg.1 ENST00000221132.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013385.hg.1 NM_003844.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013385.hg.1 TC08001052.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0800167048.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167048.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167048.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800167048.hg.1 TC0800009875.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800167048.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800167048.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167048.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000221132.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000519862.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000519862.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000519862.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000524158.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 ENST00000613472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq
          Affymetrix Human Clariom D Assay TC0800009875.hg.1 NM_003844.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-55c5c7589b-pgs97:80/100.66.130.250:80.
          git-commit: fefcbad4eb9baa7f43a2c44c49530f643471c973
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0