Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01552403_g1
          See other LIN28A GE Assays ›
          Gene Symbol:
          LIN28A
          Gene Name:
          lin-28 homolog A
          Gene Aliases:
          CSDD1, LIN-28, LIN28, ZCCHC1, lin-28A
          Chromosome Location:
          Chr.1: 26410768 - 26429728 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01552403_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LIN28A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024674.5 NP_078950.1 1 - 2 157 62
          XM_011542148.1 XP_011540450.1 1 - 2 147 62
          GenBank mRNA AF521099.1 - 1 - 2 108 62
          AK022519.1 - 1 - 2 108 62
          BC028566.2 - 1 - 2 147 3841184 62
          CN349325.1 - 1 - 2 159 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100007457.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000343.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LIN28A

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          germ cell development
          miRNA catabolic process
          stem cell population maintenance
          pre-miRNA processing
          RNA 3'-end processing
          somatic stem cell population maintenance
          positive regulation of neuron differentiation
          negative regulation of glial cell differentiation
          positive regulation of translation
          stem cell differentiation
          regulation of gene silencing by miRNA
          positive regulation of cell proliferation involved in kidney development
          DNA binding
          RNA binding
          mRNA binding
          protein binding
          zinc ion binding
          translation initiation factor binding
          miRNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01005236.hg.1 ENST00000326279.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01005236.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01005236.hg.1 TC01000343.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200010185.hg.1 ENST00000429574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01005238.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01005238.hg.1 ENST00000326279.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01005238.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01005238.hg.1 TC01000343.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1300009404.hg.1 ENST00000445283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009404.hg.1 ENST00000445283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008399.hg.1 ENST00000603272.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01005237.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01005237.hg.1 ENST00000326279.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01005237.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01005237.hg.1 TC01000343.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152064.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152064.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152064.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100152064.hg.1 TC0100007457.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100152064.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100152064.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152064.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC1300009404.hg.1 ENST00000445283.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009404.hg.1 ENST00000445283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010185.hg.1 ENST00000429574.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152061.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152061.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100152061.hg.1 TC0100007457.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100152061.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152061.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100007457.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000008399.hg.1 ENST00000603272.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152062.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152062.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152062.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100152062.hg.1 TC0100007457.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100152062.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152062.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152062.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100152063.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152063.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152063.hg.1 NM_024674.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100152063.hg.1 TC0100007457.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100152063.hg.1 ENST00000254231.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152063.hg.1 ENST00000326279.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152063.hg.1 NM_024674.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0