Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01399114_m1
          See other LINC01618 GE Assays ›
          Gene Symbol:
          LINC01618
          Gene Name:
          long intergenic non-protein coding RNA 1618
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 52789994 - 52815464 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01399114_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01618
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040106.1 4 - 5 418 94
          GenBank mRNA AL080225.1 - 4 - 5 418 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream endogenous retrovirus group MER34 member 1 ERVMER34-1 -47.9kb Anti-sense Chr. 4 - 52743517 - 52751640 on NCBI Build GRCh38
          Downstream RAS like family 11 member B RASL11B 46.9kb Sense Chr. 4 - 52862328 - 52866835 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400007518.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000302.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252465 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252467 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252464 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03098255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23252506 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3679221 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15278 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3679221 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3679221 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1564881_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR04004282.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04004282.hg.1 NR_040106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04004282.hg.1 TC04000302.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400151357.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400151357.hg.1 NR_040106.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400151357.hg.1 TC0400007518.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400151357.hg.1 NR_040106.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400151357.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 ENST00000514957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 NR_040106.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007518.hg.1 NR_040106.1 Refseq
          Human Genome U133 Plus 2.0 Array 1564881_at - -
          Human Genome U133 Plus 2.0 Array 1564881_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564881_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025517 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025517 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 508893 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1676550 NR_040106.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1676550 AL080225.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676550 NR_040106.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676550 AL080225.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1676550 NR_040106.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1676550 AL080225.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676550 NR_040106.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676550 AL080225.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1676550 NR_040106.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1676550 AL080225.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7d7666c9cf-hbrf6:80/100.66.134.79:80.
          git-commit: 173eb311d069d8bb3c8868db8478582b9fe5b4f6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0