Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01127005_m1
          See other ATP8B4 GE Assays ›
          Gene Symbol:
          ATP8B4
          Gene Name:
          ATPase phospholipid transporting 8B4 (putative)
          Gene Aliases:
          ATPIM
          Chromosome Location:
          Chr.15: 49858238 - 50182096 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01127005_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATP8B4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024837.3 NP_079113.2 7 - 8 578 90
          NR_073596.1 8 - 9 666 90
          NR_073597.1 7 - 8 578 90
          NR_073598.1 8 - 9 685 90
          XM_011522046.2 XP_011520348.1 8 - 9 883 90
          XM_011522047.2 XP_011520349.1 7 - 8 1408 90
          XM_011522048.1 XP_011520350.1 7 - 8 681 90
          XM_011522049.2 XP_011520351.1 7 - 8 791 90
          XM_011522051.2 XP_011520353.1 8 - 9 718 90
          XM_011522052.2 XP_011520354.1 7 - 8 776 90
          XM_011522053.1 XP_011520355.1 7 - 8 557 90
          XM_011522056.2 XP_011520358.2 7 - 8 943 90
          XM_011522058.2 XP_011520360.1 3 - 4 1152 90
          XM_011522059.1 XP_011520361.1 2 - 3 173 90
          XM_011522060.1 XP_011520362.1 5 - 6 408 90
          XM_011522061.1 XP_011520363.1 4 - 5 323 90
          XM_011522062.1 XP_011520364.1 2 - 3 162 90
          XM_011522063.1 XP_011520365.1 2 - 3 144 90
          XM_017022587.1 XP_016878076.1 7 - 8 943 90
          XM_017022588.1 XP_016878077.1 7 - 8 943 90
          XM_017022589.1 XP_016878078.1 7 - 8 943 90
          XM_017022590.1 XP_016878079.1 8 - 9 695 90
          XM_017022591.1 XP_016878080.1 7 - 8 642 90
          XM_017022592.1 XP_016878081.1 7 - 8 789 90
          XM_017022593.1 XP_016878082.1 7 - 8 943 90
          XM_017022594.1 XP_016878083.1 3 - 4 203 90
          XM_017022595.1 XP_016878084.1 3 - 4 2208 90
          XM_017022597.1 XP_016878086.1 7 - 8 943 90
          XM_017022598.1 XP_016878087.1 7 - 8 943 90
          XM_017022599.1 XP_016878088.1 7 - 8 943 90
          GenBank mRNA AB075819.1 - 8 - 9 685 90
          BC144297.1 - 7 - 8 574 90
          BI258773.1 - 3 - 4 217 90

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_011522060.1, XM_011522061.1, XM_011522062.1, XM_011522063.1, XM_017022595.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATP8B4

          Gene Ontology Categories:

          Function(s) Process(es)

          Golgi organization
          phospholipid translocation
          magnesium ion binding
          ATP binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0