Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01110894_s1
          See other LOC399716 GE Assays ›
          Gene Symbol:
          LOC399716
          Gene Name:
          uncharacterized LOC399716
          Gene Aliases:
          DKFZp667F0711
          Chromosome Location:
          Chr.10: 6345717 - 6352762 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01110894_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC399716
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AL713753.1 - 1 441 109

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC399715 LOC399715 -14.3kb Sense Chr. 10 - 6326544 - 6335982 on NCBI Build GRCh38
          Downstream protein kinase C theta PRKCQ PRKCT,
          nPKC-theta
          40.8kb Anti-sense Chr. 10 - 6393038 - 6580646 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047683 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16288566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047738 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402640 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402683 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047679 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047749 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402655 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16288558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047706 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047677 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402635 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402679 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047720 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047746 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402733 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047745 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16288555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047743 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402734 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16288568 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16288554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047750 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07047723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27402711 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P60606 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P60606 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 8086 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 57251 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14421 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P60606 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P60606 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02206344 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10617532 - -
          Agilent Human 1B Microarray G4111A A_32_P760092 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1559756_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1000006670.hg.1 ENST00000391437.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000006670.hg.1 ENST00000391437.2 Ensembl
          Human Genome U133 Plus 2.0 Array 1559756_at - -
          Human Genome U133 Plus 2.0 Array 1559756_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559756_at8 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 230248 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3252862 AL713753.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3178253 AL713753.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6458bd7976-5pn6j:80/100.66.131.68:80.
          git-commit: 78a20c812a412109e55a4daff1692518312115bc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/redesigning_search_header