Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015030.1 | NP_055845.1 | 62 - 63 | 9200 | 96 | |
XM_005248090.4 | XP_005248147.1 | 63 - 64 | 9258 | 96 | ||
XM_005248093.4 | XP_005248150.1 | 37 - 38 | 5885 | 96 | ||
XM_011513680.2 | XP_011511982.1 | 60 - 61 | 8717 | 96 | ||
XM_011513681.2 | XP_011511983.1 | 59 - 60 | 8702 | 96 | ||
XM_011513682.2 | XP_011511984.1 | 58 - 59 | 8657 | 96 | ||
XM_011513683.2 | XP_011511985.1 | 62 - 63 | 9094 | 96 | ||
XM_011513684.2 | XP_011511986.1 | 37 - 38 | 5891 | 96 | ||
XM_017008046.1 | XP_016863535.1 | 63 - 64 | 9525 | 96 | ||
GenBank mRNA | AB020633.1 | - | 20 - 21 | 3264 | 96 | |
AK025034.1 | - | 3 - 4 | 204 | 96 | ||
AK127307.1 | - | 24 - 25 | 3945 | 96 | ||
AK128452.1 | - | 2 - 3 | 1180 | 96 | ||
AL833170.1 | - | 28 - 29 | 4703 | 96 | ||
BC021803.1 | - | 7 - 8 | 1050 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() |