Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004756.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004756.hg.1 |
ENST00000454765.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004756.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004756.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004756.hg.1 |
TC05000345.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004754.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004754.hg.1 |
ENST00000454765.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004754.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004754.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004754.hg.1 |
TC05000345.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
TC0500007761.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152941.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
TC0500007761.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152943.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
TC0500007761.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500152944.hg.1 |
NM_173490.7 |
Refseq |