Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01077390_m1
          See other TRPC4 GE Assays ›
          Gene Symbol:
          TRPC4
          Gene Name:
          transient receptor potential cation channel subfamily C member 4
          Gene Aliases:
          HTRP-4, HTRP4, TRP4
          Chromosome Location:
          Chr.13: 37633872 - 37870374 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01077390_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRPC4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135955.1 NP_001129427.1 4 - 5 1471 79
          NM_001135956.1 NP_001129428.1 4 - 5 1471 79
          NM_001135957.1 NP_001129429.1 4 - 5 1471 79
          NM_001135958.1 NP_001129430.1 3 - 4 952 79
          NM_003306.1 NP_003297.1 4 - 5 1471 79
          NM_016179.2 NP_057263.1 4 - 5 1471 79
          GenBank mRNA AF063822.1 - 4 - 5 1471 79
          AF063823.1 - 4 - 5 1471 79
          AF063824.1 - 4 - 5 1471 79
          AF063825.1 - 4 - 5 1471 79
          AF175406.1 - 4 - 5 1467 79
          AF421358.1 - 3 - 4 1236 79
          AF421359.1 - 3 - 4 1236 79
          AF421360.1 - 3 - 4 1236 79
          AF421362.1 - 2 - 3 717 79
          AK312524.1 - 4 - 5 1441 79
          BC104725.1 - 4 - 5 1392 40008478 79
          BC105306.1 - 4 - 5 1392 79
          BC105641.1 - 4 - 5 1392 40008481 79
          BC105917.1 - 4 - 5 1392 40008477 79
          BX648312.1 - 4 - 5 2095 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300008645.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000577.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRPC4

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          manganese ion transport
          gamma-aminobutyric acid secretion
          oligodendrocyte differentiation
          regulation of cytosolic calcium ion concentration
          regulation of calcium ion transport
          calcium ion import
          calcium ion transmembrane transport
          calcium channel activity
          protein binding
          beta-catenin binding
          store-operated calcium channel activity
          cadherin binding
          inositol 1,4,5 trisphosphate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000338947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000355779.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000358477.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000379673.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000379679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000379681.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000379705.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000426868.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000447043.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 ENST00000494529.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 NM_001135955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 NM_001135956.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 NM_001135957.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 NM_001135958.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 NM_003306.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 NM_016179.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13006660.hg.1 TC13000577.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000338947.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000355779.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000358477.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000379673.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000379679.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000379705.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000426868.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000488717.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000494529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000625583.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_001135955.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_001135956.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_001135957.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_001135958.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_003306.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_016179.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 TC1300008645.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000338947.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000355779.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000358477.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000379673.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000379679.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000379705.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000426868.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000488717.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000494529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 ENST00000625583.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_001135955.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_001135956.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_001135957.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_001135958.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_003306.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300158421.hg.1 NM_016179.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0