Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145337.2 | NP_001138809.1 | 3 - 4 | 218 | 160 | |
NM_001145339.2 | NP_001138811.1 | 3 - 4 | 468 | 160 | ||
NM_002392.5 | NP_002383.2 | 3 - 4 | 468 | 160 | ||
XM_005268872.4 | XP_005268929.1 | 3 - 4 | 396 | 160 | ||
GenBank mRNA | AF092843.1 | - | 3 - 4 | 176 | 160 | |
AF092844.1 | - | 3 - 4 | 176 | 160 | ||
AF385323.1 | - | 2 - 3 | 144 | 160 | ||
AJ430612.1 | - | 2 - 3 | 144 | 160 | ||
AJ430614.1 | - | 2 - 3 | 144 | 160 | ||
AJ491698.1 | - | 2 - 3 | 144 | 160 | ||
AK290341.1 | - | 3 - 4 | 418 | 160 | ||
AM902646.1 | - | 2 - 3 | 144 | 160 | ||
AM902650.1 | - | 2 - 3 | 144 | 160 | ||
AM902651.1 | - | 2 - 3 | 144 | 160 | ||
AM902654.1 | - | 2 - 3 | 144 | 160 | ||
AM902655.1 | - | 2 - 3 | 144 | 160 | ||
AM902659.1 | - | 2 - 3 | 144 | 160 | ||
AM902666.1 | - | 2 - 3 | 144 | 160 | ||
AM902667.1 | - | 2 - 3 | 144 | 160 | ||
AM902668.1 | - | 2 - 3 | 144 | 160 | ||
AM902669.1 | - | 2 - 3 | 144 | 160 | ||
AM902672.1 | - | 2 - 3 | 144 | 160 | ||
AM902680.1 | - | 2 - 3 | 144 | 160 | ||
AM902681.1 | - | 2 - 3 | 144 | 160 | ||
BC152384.1 | - | 2 - 3 | 147 | 160 | ||
BC152390.1 | - | 2 - 3 | 147 | 160 | ||
BT007258.1 | - | 2 - 3 | 144 | 160 | ||
EU076747.1 | - | 3 - 4 | 218 | 160 | ||
EU076749.1 | - | 3 - 4 | 218 | 160 | ||
HM213901.1 | - | 2 - 3 | 144 | 160 | ||
JX015390.1 | - | 2 - 3 | 144 | 160 | ||
JX015392.1 | - | 2 - 3 | 144 | 160 | ||
M92424.1 | - | 3 - 4 | 455 | 160 | ||
Z12020.1 | - | 3 - 4 | 455 | 160 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |