Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
ENST00000413247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
ENST00000413247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
ENST00000427615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
ENST00000427615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
NR_110244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
NR_110246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
NR_110247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
TC0200009651.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
ENST00000413247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
ENST00000413247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
ENST00000427615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
ENST00000427615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
NR_110244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
NR_110246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167456.hg.1 |
NR_110247.1 |
Refseq |