Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002537.hg.1 |
ENST00000361367.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002537.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002537.hg.1 |
TC11000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000529355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000529355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000529355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000529355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000529898.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000529898.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000529898.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
ENST00000529898.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148118.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148118.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148118.hg.1 |
TC1100006840.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100148118.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148118.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000361367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000524523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000526874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000529355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000529355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000529355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000529355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000529898.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000529898.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000529898.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
ENST00000529898.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006840.hg.1 |
NM_014633.4 |
Refseq |