Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01045108_g1
          See other NDRG2 GE Assays ›
          Gene Symbol:
          NDRG2
          Gene Name:
          NDRG family member 2
          Gene Aliases:
          SYLD
          Chromosome Location:
          Chr.14: 21016763 - 21070872 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01045108_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NDRG2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282211.1 NP_001269140.1 12 - 13 867 100
          NM_001282213.1 NP_001269142.1 13 - 14 1146 100
          NM_001282214.1 NP_001269143.1 13 - 14 1134 100
          NM_001282216.1 NP_001269145.1 12 - 13 882 100
          NM_001320329.1 NP_001307258.1 13 - 14 1799 100
          NM_016250.2 NP_057334.1 13 - 14 989 100
          NM_201535.1 NP_963293.1 14 - 15 1031 100
          NM_201536.1 NP_963294.1 12 - 13 922 100
          NM_201537.1 NP_963831.1 13 - 14 964 100
          NM_201538.1 NP_963832.1 12 - 13 899 100
          NM_201539.1 NP_963833.1 13 - 14 941 100
          NM_201540.1 NP_963834.1 14 - 15 1008 100
          NM_201541.1 NP_963835.1 13 - 14 966 100
          XM_011536996.2 XP_011535298.1 14 - 15 1018 100
          XM_011536997.1 XP_011535299.1 14 - 15 1019 100
          XM_011536998.1 XP_011535300.1 14 - 15 1009 100
          XM_011536999.1 XP_011535301.1 12 - 13 1414 100
          XM_011537001.1 XP_011535303.1 13 - 14 972 100
          XM_011537002.1 XP_011535304.1 13 - 14 1118 100
          GenBank mRNA AB033074.1 - 12 - 13 851 100
          AF087872.1 - 12 - 13 905 100
          AF159092.3 - 13 - 14 912 100
          AK024521.1 - 14 - 15 964 100
          AK057843.1 - 13 - 14 935 100
          AK057847.1 - 12 - 13 899 100
          AK090657.1 - 12 - 13 838 100
          AK096999.1 - 13 - 14 1799 100
          AK098388.1 - 959 100
          AK130029.1 - 12 - 13 837 100
          AK293514.1 - 12 - 13 867 100
          AL136574.1 - 12 - 13 876 100
          BC010458.1 - 12 - 13 870 4215141 100
          BC093038.1 - 13 - 14 965 4811718 100
          BX248031.1 - 10 - 11 741 100
          BX647091.1 - 10 - 11 1064 100
          BX647748.1 - 13 - 14 1081 100
          CR749252.1 - 13 - 14 1134 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008615.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000908.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NDRG2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cytokine production
          signal transduction
          regulation of vascular endothelial growth factor production
          Wnt signaling pathway
          substantia nigra development
          cell differentiation
          negative regulation of smooth muscle cell proliferation
          negative regulation of ERK1 and ERK2 cascade
          regulation of platelet-derived growth factor production
          molecular_function
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000298684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000298687.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000350792.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000360463.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000397844.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000397847.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000397851.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000397853.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000397855.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000397856.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000397858.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000403829.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000553503.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000554104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000554143.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000554770.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000555158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 ENST00000556147.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 NM_016250.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 NM_201535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 NM_201536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 NM_201537.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 NM_201538.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 NM_201539.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 NM_201540.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 NM_201541.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009806.hg.1 TC14000908.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000298684.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000298687.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000350792.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000360463.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397847.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397851.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397853.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397856.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397858.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000403829.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000553503.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000553593.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000553793.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000554104.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000554143.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000555158.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000555650.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000556008.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000556147.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000556366.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000556716.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000557353.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000557416.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282211.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282212.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282213.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282214.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282215.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282216.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_016250.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201535.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201536.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201537.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201538.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201539.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201540.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201541.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 TC1400008615.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000360463.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000298684.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000298687.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000350792.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397847.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397851.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397853.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397856.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000397858.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000403829.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000553503.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000553593.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000553793.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000554104.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000554143.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000555158.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000555650.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000556008.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000556147.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000556366.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000556716.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000557353.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 ENST00000557416.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282211.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282212.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282213.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282214.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282215.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_001282216.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_016250.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201535.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201536.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201537.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201538.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201539.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201540.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161966.hg.1 NM_201541.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-57bdf9945f-4p2c8:80/100.66.134.79:80.
          git-commit: baa7f4fdc4fe37bd11ed6fe4edff1157ae59fd06
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0