Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019443.hg.1 |
ENST00000222823.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019443.hg.1 |
ENST00000423334.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019443.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019443.hg.1 |
TC07001249.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019445.hg.1 |
ENST00000222823.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019445.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019445.hg.1 |
TC07001249.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019442.hg.1 |
ENST00000222823.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019442.hg.1 |
ENST00000423334.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019442.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019442.hg.1 |
TC07001249.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019448.hg.1 |
ENST00000222823.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019448.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019448.hg.1 |
TC07001249.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019438.hg.1 |
ENST00000222823.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019438.hg.1 |
ENST00000423334.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019438.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019438.hg.1 |
TC07001249.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000411552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000419799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000434755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
TC0700010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000411552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000419799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
ENST00000434755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175600.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175604.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175604.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175604.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175604.hg.1 |
TC0700010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175604.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175604.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175604.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000411552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000419799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000434755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
TC0700010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000411552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000419799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
ENST00000434755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175601.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000411552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000413433.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000419799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000434755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
TC0700010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000411552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000413433.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000419799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
ENST00000434755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175595.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000411552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000413433.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000419799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000434755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
NM_006092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
TC0700010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000222823.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000411552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000413433.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000419601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000419799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
ENST00000434755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175599.hg.1 |
NM_006092.3 |
Refseq |