Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023727.hg.1 |
ENST00000367649.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023727.hg.1 |
ENST00000448150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023727.hg.1 |
NM_170692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023727.hg.1 |
TC01001539.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023729.hg.1 |
ENST00000367649.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023729.hg.1 |
ENST00000448150.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023729.hg.1 |
ENST00000462775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023729.hg.1 |
NM_004841.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023729.hg.1 |
NM_170692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023729.hg.1 |
TC01001539.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100178842.hg.1 |
ENST00000367649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178842.hg.1 |
NM_170692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178842.hg.1 |
TC0100010735.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100178842.hg.1 |
NM_170692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178842.hg.1 |
ENST00000367649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
ENST00000367649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
ENST00000462775.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
NM_004841.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
NM_170692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
TC0100010735.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
ENST00000367649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
ENST00000462775.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
NM_004841.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178845.hg.1 |
NM_170692.2 |
Refseq |