Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000498745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000498745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164615.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164615.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164615.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164615.hg.1 |
TC0700009079.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700164615.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164615.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164615.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000486685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000496963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000498745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000498745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
ENST00000614309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009079.hg.1 |
NM_178562.4 |
Refseq |