Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004191.hg.1 |
ENST00000261716.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004191.hg.1 |
ENST00000536202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004191.hg.1 |
NM_020791.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004191.hg.1 |
NM_025142.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004191.hg.1 |
TC17000341.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
ENST00000261716.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
ENST00000536202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
ENST00000578653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
NM_020791.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
NM_025142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
NM_020791.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
TC1700007419.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
ENST00000261716.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
ENST00000536202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
ENST00000578653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152900.hg.1 |
NM_025142.1 |
Refseq |