Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002241.hg.1 |
TC22000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002241.hg.1 |
ENST00000263116.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002241.hg.1 |
ENST00000341989.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002241.hg.1 |
ENST00000418881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002241.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002239.hg.1 |
TC22000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002239.hg.1 |
ENST00000263116.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002239.hg.1 |
ENST00000341989.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002239.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002240.hg.1 |
TC22000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002240.hg.1 |
ENST00000263116.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002240.hg.1 |
ENST00000341989.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002240.hg.1 |
ENST00000418881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002240.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002238.hg.1 |
TC22000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002238.hg.1 |
ENST00000263116.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002238.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
ENST00000420895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006832.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200147008.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147008.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200147008.hg.1 |
TC2200006832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200147008.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200147008.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147010.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147010.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147010.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200147010.hg.1 |
TC2200006832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200147010.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147010.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147010.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200147009.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147009.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147009.hg.1 |
NM_004914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200147009.hg.1 |
TC2200006832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200147009.hg.1 |
ENST00000263116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147009.hg.1 |
ENST00000341989.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147009.hg.1 |
NM_004914.4 |
Refseq |