Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01008539_s1
          See other FLJ31713 GE Assays ›
          Gene Symbol:
          FLJ31713
          Gene Name:
          uncharacterized protein FLJ31713
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01008539_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ31713
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK056275.1 - 1 2648 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB323823.1 - 1 Chr. 9 - 113614589 - 113615117 [+] on Build GRCh38 108

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream regulator of G-protein signaling 3 RGS3 C2PA,
          RGP3
          -16.8kb Sense Chr. 9 - 113444729 - 113597743 on NCBI Build GRCh38
          Downstream zinc finger protein 618 ZNF618 FP13169,
          NEDD10
          261.2kb Sense Chr. 9 - 113876282 - 114056595 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16255471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16255476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908129 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908119 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16255472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908089 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908099 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908085 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27252211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16255473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06908090 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111476 AK090672.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111476 BC100814.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111476 BC100815.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111476 BC100816.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111476 BC100817.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111476 NM_177402.3 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CGH Microarray 8x60K 10250 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 137831 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3318327 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P354326 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 31389 - -
          Agilent Human Genome CGH Microarray 44B A_14_P112258 - -
          Agilent Human Genome CGH Microarray 44A A_14_P112258 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59515 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P112258 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3318327 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P354326 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P112258 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P112258 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P112258 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P354326 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 31550 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 98860 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P112258 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P112258 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_041881_at - -
          Affymetrix Human Gene 1.0 ST Array XR_041883_at - -
          Affymetrix Human Gene 1.0 ST Array XR_041882_at - -
          Affymetrix Human Gene 1.0 ST Array 8163534 - -
          Affymetrix Human Gene 1.1 ST Array 8163533 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553429_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553429_at - -
          Human Genome U133 Plus 2.0 Array 1553429_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553429_at1 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-98ffbb479-9qks8:80/100.66.134.79:80.
          git-commit: cc4aab6ff2cd04db3b50ee2b2ac7bb13b20eb648
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0