Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01005512_g1
          See other KMT2C GE Assays ›
          Gene Symbol:
          KMT2C
          Gene Name:
          lysine methyltransferase 2C
          Gene Aliases:
          HALR, MLL3
          Chromosome Location:
          Chr.7: 152134925 - 152436642 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01005512_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KMT2C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_170606.2 NP_733751.2 30 - 31 4759 95
          XM_005250025.4 XP_005250082.1 32 - 33 4613 95
          XM_005250026.3 XP_005250083.1 32 - 33 4647 95
          XM_005250027.4 XP_005250084.1 31 - 32 4821 95
          XM_005250028.4 XP_005250085.1 31 - 32 4821 95
          XM_005250031.4 XP_005250088.1 31 - 32 4821 95
          XM_006716077.3 XP_006716140.1 31 - 32 4821 95
          XM_006716078.3 XP_006716141.1 31 - 32 4821 95
          XM_006716079.3 XP_006716142.1 31 - 32 4821 95
          XM_011516450.2 XP_011514752.1 30 - 31 4773 95
          XM_011516451.2 XP_011514753.1 30 - 31 4701 95
          XM_011516452.2 XP_011514754.1 29 - 30 4668 95
          XM_011516453.2 XP_011514755.1 31 - 32 4821 95
          XM_011516454.2 XP_011514756.1 26 - 27 3816 95
          XM_011516456.2 XP_011514758.1 30 - 31 4773 95
          XM_017012480.1 XP_016867969.1 32 - 33 4650 95
          XM_017012481.1 XP_016867970.1 32 - 33 4610 95
          XM_017012482.1 XP_016867971.1 31 - 32 4821 95
          XM_017012483.1 XP_016867972.1 31 - 32 4821 95
          XM_017012484.1 XP_016867973.1 31 - 32 4788 95
          XM_017012485.1 XP_016867974.1 30 - 31 4770 95
          XM_017012486.1 XP_016867975.1 31 - 32 4821 95
          XM_017012487.1 XP_016867976.1 31 - 32 4789 95
          XM_017012488.1 XP_016867977.1 31 - 32 4821 95
          XM_017012489.1 XP_016867978.1 12 - 13 1328 95
          XM_017012490.1 XP_016867979.1 9 - 10 900 95
          GenBank mRNA AB040939.2 - 19 - 20 2875 95
          AF264750.2 - 16 - 17 2086 95
          AY024361.1 - 30 - 31 4720 95
          BC151837.1 - 19 - 20 2875 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700013102.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07002045.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KMT2C

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3-K4 methylation
          DNA binding
          RNA binding
          protein binding
          zinc ion binding
          acyltransferase activity
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07031383.hg.1 ENST00000262189.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031383.hg.1 ENST00000355193.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031383.hg.1 NM_170606.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07031383.hg.1 TC07002045.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 ENST00000262189.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 ENST00000355193.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 ENST00000473186.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 ENST00000558084.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 NM_170606.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 TC0700013102.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 ENST00000262189.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 ENST00000355193.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 ENST00000473186.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 ENST00000558084.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194595.hg.1 NM_170606.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0