Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01004341_m1
          See other RAB14 GE Assays ›
          Gene Symbol:
          RAB14
          Gene Name:
          RAB14, member RAS oncogene family
          Gene Aliases:
          FBP, RAB-14
          Chromosome Location:
          Chr.9: 121178137 - 121202087 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01004341_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RAB14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016322.3 NP_057406.2 1 - 2 449 115
          GenBank mRNA AK023524.1 - 1 - 2 231 115
          AK090889.1 - 1 - 2 248 115
          AL162081.1 - 1 - 2 176 115
          BC006081.1 - 1 - 2 220 2963119 115
          DA404780.1 - 1 - 2 449 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011396.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001543.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK023524.1, AK090889.1, AL162081.1, BC006081.1, NM_016322.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RAB14

          Gene Ontology Categories:

          Function(s) Process(es)

          Golgi to endosome transport
          small GTPase mediated signal transduction
          body fluid secretion
          fibroblast growth factor receptor signaling pathway
          embryo development
          protein transport
          vesicle-mediated transport
          endocytic recycling
          regulation of protein localization
          defense response to bacterium
          apical protein localization
          intracellular transport
          membrane organization
          phagolysosome assembly involved in apoptotic cell clearance
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          myosin V binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09019449.hg.1 ENST00000373840.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019449.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019449.hg.1 TC09001543.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09019450.hg.1 ENST00000373840.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019450.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019450.hg.1 TC09001543.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09019452.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019452.hg.1 TC09001543.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900177763.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900177763.hg.1 TC0900011396.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177763.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900177761.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177761.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900177761.hg.1 TC0900011396.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177761.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177761.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011396.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900177759.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177759.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177759.hg.1 NM_016322.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900177759.hg.1 TC0900011396.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177759.hg.1 ENST00000373840.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177759.hg.1 ENST00000451303.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177759.hg.1 NM_016322.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 145778 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0