Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00995149_m1
          See other B4GALT1 GE Assays ›
          Gene Symbol:
          B4GALT1
          Gene Name:
          beta-1,4-galactosyltransferase 1
          Gene Aliases:
          B4GAL-T1, CDG2D, GGTB2, GT1, GTB, beta4Gal-T1
          Chromosome Location:
          Chr.9: 33110641 - 33167391 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00995149_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B4GALT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001497.3 NP_001488.2 3 - 4 1026 74
          GenBank mRNA A23697.1 - 3 - 4 843 74
          AK296928.1 - 4 - 5 725 74
          AK297541.1 - 2 - 3 360 74
          AK312797.1 - 3 - 4 865 74
          AK316294.1 - 4 - 5 698 74
          D29805.1 - 3 - 4 909 74
          M13701.1 - 2 - 3 426 74
          M22921.1 - 3 - 4 909 74
          U10472.1 - 1 - 2 62 74
          U10473.1 - 3 - 4 567 74
          U10474.1 - 3 - 4 910 74
          X13223.1 - 3 - 4 909 74
          X14085.1 - 3 - 4 906 74
          X55415.1 - 3 - 4 837 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900009846.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001008.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B4GALT1

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial cell development
          acute inflammatory response
          lactose biosynthetic process
          galactose metabolic process
          protein N-linked glycosylation
          cell adhesion
          binding of sperm to zona pellucida
          penetration of zona pellucida
          negative regulation of cell population proliferation
          oligosaccharide biosynthetic process
          keratan sulfate biosynthetic process
          extracellular matrix organization
          mammary gland development
          development of secondary sexual characteristics
          branching morphogenesis of an epithelial tube
          leukocyte migration
          regulation of cellular component movement
          regulation of acrosome reaction
          positive regulation of epithelial cell proliferation involved in wound healing
          angiogenesis involved in wound healing
          positive regulation of apoptotic process involved in mammary gland involution
          beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase activity
          N-acetyllactosamine synthase activity
          lactose synthase activity
          cytoskeletal protein binding
          galactosyltransferase activity
          protein kinase binding
          manganese ion binding
          UDP-galactosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09013211.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09013211.hg.1 ENST00000541701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09013211.hg.1 ENST00000541851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09013211.hg.1 NM_001497.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09013211.hg.1 TC09001008.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000535206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000535206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000535206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 ENST00000535206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900167366.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900167366.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900167366.hg.1 TC0900009846.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900167366.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900167366.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000379731.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000535206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000535206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000535206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 ENST00000535206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 NM_001497.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009846.hg.1 NM_001497.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0