Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00994679_s1
          See other JUND GE Assays ›
          Gene Symbol:
          JUND
          Gene Name:
          JunD proto-oncogene, AP-1 transcription factor subunit
          Gene Aliases:
          AP-1
          Chromosome Location:
          Chr.19: 18279694 - 18281656 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00994679_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: JUND
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286968.1 NP_001273897.1 1 1915 99
          NM_005354.5 NP_005345.3 1 1915 99
          GenBank mRNA AW027337.1 - 1 425 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900010009.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene JUND

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          osteoblast development
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          aging
          circadian rhythm
          response to radiation
          response to xenobiotic stimulus
          response to light stimulus
          response to mechanical stimulus
          regulation of cell death
          response to lipopolysaccharide
          cellular response to hormone stimulus
          response to cytokine
          regulation of cell population proliferation
          response to peptide hormone
          positive regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of transcription by RNA polymerase II
          response to cAMP
          regulation of cell cycle
          cellular response to calcium ion
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          transcription coactivator activity
          protein binding
          transcription factor binding
          nuclear receptor binding
          enzyme binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 ENST00000600972.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 ENST00000600972.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900182195.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900182195.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900182195.hg.1 TC1900010009.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900182195.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900182195.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 ENST00000600972.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 ENST00000600972.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay TC1900010009.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 ENST00000600972.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 TC1900010009.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 ENST00000600972.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900182197.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900182196.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900182196.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900182196.hg.1 NM_005354.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900182196.hg.1 TC1900010009.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900182196.hg.1 ENST00000252818.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900182196.hg.1 NM_001286968.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900182196.hg.1 NM_005354.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 165338 - -
          ABI Human Genome Survey Microarray Version 2.0 230932 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0