Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00991397_s1
          See other FLJ21369 GE Assays ›
          Gene Symbol:
          FLJ21369
          Gene Name:
          uncharacterized protein FLJ21369
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00991397_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ21369
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK025022.1 - 1 1547 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LSM14A,
          mRNA processing body assembly factor
          LSM14A C19orf13,
          FAM61A,
          RAP55,
          RAP55A
          -46.9kb Sense Chr. 19 - 34172428 - 34229515 on NCBI Build GRCh38
          Downstream glucose-6-phosphate isomerase GPI AMF,
          GNPI,
          NLK,
          PGI,
          PHI,
          SA-36,
          SA36
          81.1kb Sense Chr. 19 - 34359592 - 34402413 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3301752 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P50597 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 35855 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42249 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3301752 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3301752 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P50597 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P50597 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 220401_PM_at - -
          Human Genome U133 Plus 2.0 Array 220401_PM_at - -
          Human Genome U133 Plus 2.0 Array 220401_at - -
          Human Genome U133A 2.0 Array 220401_at - -
          Human Genome U133A 2.0 Array 220401_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw518807 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220401_at10 - -
          Human Genome U95 Set 83905_at - -
          Human Genome U95 Set 83905_PM_at - -
          Human Genome U133 Set 220401_at - -
          Human Genome U133 Set 220401_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0