Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00983251_g1
          See other MAP2K1 GE Assays ›
          Gene Symbol:
          MAP2K1
          Gene Name:
          mitogen-activated protein kinase kinase 1
          Gene Aliases:
          CFC3, MAPKK1, MEK1, MKK1, PRKMK1
          Chromosome Location:
          Chr.15: 66386873 - 66491544 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00983251_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAP2K1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002755.3 NP_002746.1 4 - 5 988 97
          XM_011521783.2 XP_011520085.1 4 - 5 645 97
          GenBank mRNA AK291500.1 - 4 - 5 949 97
          AK294322.1 - 4 - 5 499 97
          AK312356.1 - 4 - 5 895 97
          BC137459.1 - 4 - 5 576 9021459 97
          BC139729.1 - 4 - 5 572 40053655 97
          L05624.1 - 4 - 5 546 97
          L11284.1 - 4 - 5 585 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500007614.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000613.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAP2K1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          movement of cell or subcellular component
          chemotaxis
          signal transduction
          heart development
          negative regulation of cell population proliferation
          positive regulation of gene expression
          negative regulation of gene expression
          peptidyl-tyrosine phosphorylation
          cerebellar cortex formation
          neuron differentiation
          keratinocyte differentiation
          thyroid gland development
          regulation of stress-activated MAPK cascade
          positive regulation of transcription, DNA-templated
          thymus development
          regulation of axon regeneration
          cell motility
          positive regulation of axonogenesis
          Bergmann glial cell differentiation
          face development
          trachea formation
          epithelial cell proliferation involved in lung morphogenesis
          placenta blood vessel development
          labyrinthine layer development
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of protein serine/threonine kinase activity
          regulation of Golgi inheritance
          cellular senescence
          positive regulation of mitophagy in response to mitochondrial depolarization
          positive regulation of production of miRNAs involved in gene silencing by miRNA
          regulation of early endosome to late endosome transport
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein tyrosine kinase activity
          protein binding
          ATP binding
          protein C-terminus binding
          protein kinase binding
          protein serine/threonine kinase activator activity
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15005020.hg.1 ENST00000307102.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15005020.hg.1 NM_002755.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15005020.hg.1 TC15000613.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500153981.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153981.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153981.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500153981.hg.1 TC1500007614.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500153981.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500153981.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153981.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000307102.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000425818.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 ENST00000566326.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007614.hg.1 NM_002755.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-69f987f899-9tng4:80/100.66.130.250:80.
          git-commit: eb1b8c2b8eec225754209f035b16fef014668c1b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0