Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000407030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011392.hg.1 |
ENST00000611928.1 |
Ensembl |