Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000487569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000487569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000487569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
ENST00000487569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003726.hg.1 |
ENST00000207870.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003726.hg.1 |
ENST00000472721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003726.hg.1 |
ENST00000542835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003726.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003726.hg.1 |
TC03000191.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0300203693.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203693.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203693.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203693.hg.1 |
TC0300013807.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300203693.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203693.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203693.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000207870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000424034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000427323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000487569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000487569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000487569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
ENST00000487569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013807.hg.1 |
NM_005108.3 |
Refseq |