Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011638.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011638.hg.1 |
ENST00000410021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011638.hg.1 |
ENST00000541757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011638.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011638.hg.1 |
TC02000749.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000412481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000412481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000412481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000412481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000494979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000494979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000494979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
ENST00000494979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163373.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163373.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163373.hg.1 |
TC0200009055.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200163373.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163373.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000327097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000412481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000412481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000412481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000412481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000494979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000494979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000494979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
ENST00000494979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009055.hg.1 |
NM_006770.3 |
Refseq |