Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017539.2 | NP_060009.1 | 40 - 41 | 5774 | 92 | |
XM_006721062.1 | XP_006721125.1 | 40 - 41 | 5912 | 92 | ||
XM_011545882.2 | XP_011544184.1 | 41 - 42 | 6352 | 92 | ||
XM_011545883.1 | XP_011544185.1 | 41 - 42 | 6120 | 92 | ||
XM_011545885.2 | XP_011544187.1 | 41 - 42 | 6352 | 92 | ||
XM_011545886.2 | XP_011544188.1 | 33 - 34 | 4727 | 92 | ||
XM_011545888.2 | XP_011544190.1 | 23 - 24 | 3171 | 92 | ||
XM_011545889.2 | XP_011544191.1 | 23 - 24 | 3135 | 92 | ||
XM_017023426.1 | XP_016878915.1 | 40 - 41 | 6217 | 92 | ||
XM_017023427.1 | XP_016878916.1 | 39 - 40 | 6109 | 92 | ||
XM_017023428.1 | XP_016878917.1 | 41 - 42 | 6352 | 92 | ||
XM_017023429.1 | XP_016878918.1 | 41 - 42 | 6353 | 92 | ||
XM_017023432.1 | XP_016878921.1 | 40 - 41 | 5912 | 92 | ||
GenBank mRNA | AF494040.1 | - | 40 - 41 | 5774 | 92 | |
AK125907.1 | - | 17 - 18 | 2491 | 92 |
Set Membership: |
![]() ![]() ![]() ![]() |