Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00952035_mH
          See other CTSV GE Assays ›
          Gene Symbol:
          CTSV
          Gene Name:
          cathepsin V
          Gene Aliases:
          CATL2, CTSL2, CTSU
          Chromosome Location:
          Chr.9: 97029677 - 97039643 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00952035_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTSV
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001201575.1 NP_001188504.1 5 - 6 674 88
          NM_001333.3 NP_001324.2 5 - 6 877 88
          GenBank mRNA AB001928.1 - 5 - 6 700 88
          AF070448.1 - 4 - 5 654 88
          AK310740.1 - 5 - 6 809 88
          AK312806.1 - 5 - 6 674 88
          AY358641.1 - 5 - 6 877 88
          BC023504.2 - 5 - 6 691 3139314 88
          BC110512.1 - 5 - 6 746 40031744 88
          Y14734.1 - 5 - 6 696 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900010933.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001381.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CTSV

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          cellular response to starvation
          response to glucose
          multicellular organism aging
          negative regulation of keratinocyte proliferation
          protein autoprocessing
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          nerve development
          extracellular matrix disassembly
          hair follicle morphogenesis
          response to gonadotropin
          decidualization
          autophagic cell death
          response to glucocorticoid
          proteolysis involved in cellular protein catabolic process
          Sertoli cell differentiation
          regulation of actin cytoskeleton reorganization
          aminopeptidase activity
          cysteine-type endopeptidase activity
          serine-type endopeptidase activity
          protein binding
          cysteine-type peptidase activity
          cysteine-type carboxypeptidase activity
          kininogen binding
          peptide binding
          histone binding
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09017278.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017278.hg.1 ENST00000538255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017278.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017278.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017278.hg.1 TC09001381.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09017280.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017280.hg.1 ENST00000538255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017280.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017280.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017280.hg.1 TC09001381.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09017292.hg.1 ENST00000538255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017292.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017292.hg.1 TC09001381.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017281.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017281.hg.1 ENST00000538255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017281.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017281.hg.1 NM_001333.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017281.hg.1 TC09001381.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 TC0900010933.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174410.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 TC0900010933.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174409.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174420.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174420.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174420.hg.1 TC0900010933.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174420.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174420.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000479932.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010933.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 NM_001333.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 TC0900010933.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 ENST00000259470.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 ENST00000538255.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 NM_001201575.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174408.hg.1 NM_001333.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6dd4f99df8-bsnbs:80/100.66.135.8:80.
          git-commit: 33aac1f801e405ffbac6f281a4ca9f2baedc1304
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0