Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006057.hg.1 |
ENST00000287218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006057.hg.1 |
ENST00000373391.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006057.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006057.hg.1 |
TC06000549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000469208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000469208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000469208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
TC0600007887.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000469208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154807.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000287218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000373391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000440482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000463847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000469208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000469208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
ENST00000474522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007887.hg.1 |
NM_021943.2 |
Refseq |