Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459494 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638443 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638402 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638425 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459532 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638434 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459471 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459535 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638412 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15762130 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638454 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638408 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459479 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638407 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459525 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638461 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638445 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459482 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459538 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638383 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638446 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459461 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15762131 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459500 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638392 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638428 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459456 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459465 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15762132 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459455 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638384 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459545 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638436 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459510 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459511 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638389 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459517 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638416 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638414 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15762128 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459463 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638423 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459531 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638453 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459541 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459487 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638411 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459483 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459514 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638447 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459542 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638426 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638440 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459468 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459496 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459470 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459519 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638401 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459457 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459524 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459498 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459480 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638377 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638419 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459452 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459493 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459513 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638441 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459473 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459518 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459464 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459512 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24638375 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04459530 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
697349 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
229434 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
537548 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
233377 |
AB018299.3 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
233377 |
AK091338.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
233377 |
AX746920.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
233377 |
BC035389.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
233377 |
NM_015090.2 |
refseq_mrna_human_NM |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P922631 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_24_P922631 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
10470 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
22047 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31641499 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P01393289 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P37461736 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P922631 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_24_P922631 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459471 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638392 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459500 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638400 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638419 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459461 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638443 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459480 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638411 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638416 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638423 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459487 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459464 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459525 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638408 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638428 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459519 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638401 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459509 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459514 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638414 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638441 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638399 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638445 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459473 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638398 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459468 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459458 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459479 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638389 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459465 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638412 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638420 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638440 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459496 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459493 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459517 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638422 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459498 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638384 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638396 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459513 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638382 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459518 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638424 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638426 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638402 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459483 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459471 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459464 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638426 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459468 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459498 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459465 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638422 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638389 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459480 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459517 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638408 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638401 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638382 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459519 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638399 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638428 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459509 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638443 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638416 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638445 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459514 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459487 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459525 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638440 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638423 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638420 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459513 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459479 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638402 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459461 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459483 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638392 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638419 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638412 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459493 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459458 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638411 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638384 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638398 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638424 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459473 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459500 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459518 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638414 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638400 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04459496 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638441 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24638396 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_24_P922631 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_24_P922631 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P24691018 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P15495322 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P37461722 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P17395060 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P24691348 |
- |
- |
Agilent Human 1B Microarray G4111A |
A_32_P372388 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459464 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459539 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459518 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459508 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459541 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459513 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459490 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459514 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459469 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459505 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459530 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459471 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459528 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459492 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459537 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459494 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459503 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459473 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459496 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459456 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459501 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459516 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459499 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459520 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459453 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459480 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459544 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459543 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459466 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459509 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459522 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459475 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459507 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459476 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15762131 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459458 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459504 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459526 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459533 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459483 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459536 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459461 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04459487 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P37461734 |
- |
- |