Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013811.hg.1 |
ENST00000378643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013811.hg.1 |
ENST00000543657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013811.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013811.hg.1 |
TC09001049.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168153.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168153.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168153.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168153.hg.1 |
TC0900009930.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900168153.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168153.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168153.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |