Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00909568_g1
          See other ATF4 GE Assays ›
          Gene Symbol:
          ATF4
          Gene Name:
          activating transcription factor 4
          Gene Aliases:
          CREB-2, CREB2, TAXREB67, TXREB
          Chromosome Location:
          Chr.22: 39519709 - 39522686 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00909568_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATF4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182810.2 NP_877962.1 1 - 2 188 103
          XM_017028807.1 XP_016884296.1 2 - 3 667 103
          GenBank mRNA AK057751.1 - 1 - 2 192 103
          AK296028.1 - 1 - 2 194 103
          BC008090.1 - 1 - 2 172 3454473 103
          BC011994.1 - 1 - 2 170 3882614 103
          BC016855.2 - 1 - 2 164 3850361 103
          BC022088.2 - 1 - 2 171 3030097 103
          BC024775.1 - 1 - 2 170 4992862 103
          BC044895.1 - 1 - 2 156 4513828 103
          BC073754.1 - 1 - 2 161 6580199 103
          BC073990.1 - 1 - 2 136 6022488 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007406.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000317.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK296028.1, BC008090.1, BC011994.1, BC016855.2, BC022088.2, BC024775.1, BC044895.1, BC073754.1, BC073990.1, NM_182810.2, XM_017028807.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human CMV and MAPK Pathways Human NGF Pathway Human Interferon Pathway Human Toll-Like Receptors Pathway Human BMP Pathway HUMAN CREB PATHWAY HUMAN INTRACELLULAR CALCIUM SIGNALING HUMAN PKA SIGNALING HUMAN GPCR PATHWAY HUMAN IGF1R SIGNALING 1700 Inventoried Human_Toll-Like_Receptors_Pathway Human_Interferon_Pathway Human_BMP_Pathway Human_NGF_Pathway Human_Intracellular_Calcium_Signaling Human_GPCR_Pathway Human_PKA_Signaling Human_CREB_Pathway Human_IGF1R_Signaling Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATF4

          Gene Ontology Categories:

          Function(s) Process(es)

          gluconeogenesis
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          cellular amino acid metabolic process
          gamma-aminobutyric acid signaling pathway
          positive regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          negative regulation of translational initiation in response to stress
          circadian regulation of gene expression
          cellular response to amino acid starvation
          cellular response to UV
          response to endoplasmic reticulum stress
          positive regulation of transcription from RNA polymerase II promoter in response to stress
          positive regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          PERK-mediated unfolded protein response
          cellular response to glucose starvation
          mRNA transcription by RNA polymerase II
          positive regulation of apoptotic process
          negative regulation of potassium ion transport
          positive regulation of neuron apoptotic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase I
          positive regulation of transcription by RNA polymerase II
          positive regulation of transcription from RNA polymerase II promoter in response to arsenic-containing substance
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          negative regulation of oxidative stress-induced neuron death
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          response to manganese-induced endoplasmic reticulum stress
          transcription cis-regulatory region binding
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          core promoter sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          protein C-terminus binding
          leucine zipper domain binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00008847.hg.1 ENST00000601944.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009092.hg.1 ENST00000393544.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009092.hg.1 ENST00000393544.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009092.hg.1 ENST00000393544.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009092.hg.1 ENST00000438498.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009092.hg.1 ENST00000438498.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009092.hg.1 ENST00000544730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22006022.hg.1 TC22000317.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22006022.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006022.hg.1 ENST00000396680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006022.hg.1 ENST00000404241.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006022.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22006022.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011215.hg.1 ENST00000443279.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006008.hg.1 TC22000317.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22006008.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006008.hg.1 ENST00000396680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006008.hg.1 ENST00000404241.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006008.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22006008.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22006010.hg.1 TC22000317.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22006010.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006010.hg.1 ENST00000396680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006010.hg.1 ENST00000404241.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006010.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22006010.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22006021.hg.1 TC22000317.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22006021.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006021.hg.1 ENST00000396680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006021.hg.1 ENST00000404241.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006021.hg.1 NM_001675.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22006021.hg.1 NM_182810.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011789.hg.1 ENST00000433309.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 TC2200007406.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152252.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 TC2200007406.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200152251.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay TC1700011789.hg.1 ENST00000433309.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 TC2200007406.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200152249.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008847.hg.1 ENST00000601944.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011215.hg.1 ENST00000443279.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009092.hg.1 ENST00000393544.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009092.hg.1 ENST00000393544.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009092.hg.1 ENST00000393544.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009092.hg.1 ENST00000438498.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009092.hg.1 ENST00000438498.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009092.hg.1 ENST00000544730.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000337304.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000396680.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 ENST00000404241.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_001675.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_182810.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007406.hg.1 NM_182810.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 183867 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0