Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000487234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000487234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000487234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000468704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000480937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000487234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000487234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000487234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000491936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000498135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
NM_014313.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007398.hg.1 |
ENST00000374358.4 |
Ensembl |