Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00900501_m1
          See other PELI1 GE Assays ›
          Gene Symbol:
          PELI1
          Gene Name:
          pellino E3 ubiquitin protein ligase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 64092652 - 64144471 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00900501_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PELI1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020651.3 NP_065702.2 2 - 3 531 118
          XM_011532994.2 XP_011531296.1 2 - 3 1190 118
          XM_017004520.1 XP_016860009.1 3 - 4 665 118
          XM_017004521.1 XP_016860010.1 2 - 3 480 118
          GenBank mRNA AF300987.1 - 1 - 2 86 118
          AK022261.1 - 2 - 3 422 118
          AK312441.1 - 2 - 3 480 118
          BC050019.1 - 2 - 3 485 5296990 118
          BC050533.1 - 2 - 3 467 118
          BC063611.1 - 2 - 3 459 4992160 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200012809.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001911.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF300987.1, AK312441.1, BC050019.1, BC063611.1, NM_020651.3, XM_011532994.2, XM_017004520.1, XM_017004521.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PELI1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine production
          Toll signaling pathway
          positive regulation of B cell proliferation
          positive regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          response to lipopolysaccharide
          positive regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          negative regulation of T cell proliferation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to dsRNA
          protein K48-linked ubiquitination
          protein binding
          ligase activity
          ubiquitin-ubiquitin ligase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02030572.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030572.hg.1 ENST00000466177.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030572.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030572.hg.1 TC02001911.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02030580.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030580.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030580.hg.1 TC02001911.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030579.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030579.hg.1 ENST00000466177.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030579.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030579.hg.1 TC02001911.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02030576.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030576.hg.1 ENST00000466177.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030576.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030576.hg.1 TC02001911.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02030584.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030584.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030584.hg.1 TC02001911.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 TC0200012809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193438.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193436.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193436.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193436.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193436.hg.1 TC0200012809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193436.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193436.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193436.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 TC0200012809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193443.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 TC0200012809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193440.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 TC0200012809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193439.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193448.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193448.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193448.hg.1 TC0200012809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193448.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193448.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 TC0200012809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193444.hg.1 NM_020651.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 113197 - -
          ABI Human Genome Survey Microarray Version 2.0 207395 - -
          ABI Human Genome Survey Microarray Version 2.0 222128 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0