Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00899846_m1
          See other IL1A GE Assays ›
          Gene Symbol:
          IL1A
          Gene Name:
          interleukin 1 alpha
          Gene Aliases:
          IL-1A, IL1, IL1-ALPHA, IL1F1
          Chromosome Location:
          Chr.2: 112773915 - 112785398 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00899846_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IL1A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000575.4 NP_000566.3 3 - 4 1056 115
          GenBank mRNA AK314850.1 - 3 - 4 151 115
          BC013142.2 - 2 - 3 101 3875617 115
          BT007014.1 - 2 - 3 92 115
          CR457414.1 - 2 - 3 92 115
          M15329.1 - 2 - 3 97 115
          X02531.1 - 2 - 3 92 115
          X02851.1 - 3 - 4 150 115
          X56086.1 - 3 - 4 118 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013913.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002218.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IL1A

          Gene Ontology Categories:

          Function(s) Process(es)

          fever generation
          positive regulation of cytokine production
          connective tissue replacement involved in inflammatory response wound healing
          apoptotic process
          inflammatory response
          immune response
          cell population proliferation
          negative regulation of cell population proliferation
          positive regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          cytokine-mediated signaling pathway
          cellular response to heat
          ectopic germ cell programmed cell death
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of angiogenesis
          positive regulation of mitotic nuclear division
          positive regulation of transcription by RNA polymerase II
          positive regulation of JNK cascade
          response to copper ion
          positive regulation of protein secretion
          positive regulation of cell division
          extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          cytokine activity
          interleukin-1 receptor binding
          copper ion binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02036036.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02036036.hg.1 NM_000575.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02036036.hg.1 TC02002218.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200202060.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200202060.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200202060.hg.1 TC0200013913.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200202060.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200202060.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 ENST00000263339.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq
          Affymetrix Human Clariom D Assay TC0200013913.hg.1 NM_000575.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0