Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012225.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012225.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012225.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012225.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012225.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012225.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012225.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012225.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184142.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184142.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184142.hg.1 |
TC0600012225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600184142.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184142.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184144.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184144.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184144.hg.1 |
TC0600012225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600184144.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184144.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012225.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012225.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012225.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012225.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012225.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012225.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012225.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012225.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184143.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184143.hg.1 |
NR_125854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184143.hg.1 |
TC0600012225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600184143.hg.1 |
ENST00000445346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184143.hg.1 |
NR_125854.1 |
Refseq |