Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00540889_m1
          See other LINC00315 GE Assays ›
          Gene Symbol:
          LINC00315
          Gene Name:
          long intergenic non-protein coding RNA 315
          Gene Aliases:
          C21orf93, NCRNA00315
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00540889_m1
          Size
          Availability Made To Order
          Catalog #
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00315
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF427488.1 - 1 - 2 769 88

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF427488.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079106 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079074 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17243446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17243438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17243445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32079047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11416237 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 41369 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18558 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P100190 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 170890 BC006356.2 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 170890 BC006396.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 170890 BC018611.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 170890 NM_016170.3 refseq_mrna_human_NM
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 94047 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P131234 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596537 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596534 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596530 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8070898 - -
          Affymetrix Human Gene 1.0 ST Array 8070899 - -
          Affymetrix Human Gene 1.0 ST Array 8070897 - -
          Affymetrix Human Gene 1.1 ST Array 8070896 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553832_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2100008410.hg.1 ENST00000441947.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008410.hg.1 ENST00000441947.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008410.hg.1 ENST00000441947.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008410.hg.1 ENST00000441947.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008410.hg.1 ENST00000441947.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008410.hg.1 ENST00000441947.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1553832_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553832_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553832_at10 - -
          Human Genome U95 Set 85453_PM_at - -
          Human Genome U95 Set 85453_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 225083 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1752312 AF427488.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1752312 AF427488.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1752312 AF427488.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1752312 AF427488.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1752312 AF427488.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5f68d85665-v9rd4:80/100.66.130.250:80.
          git-commit: a5c360236fb87e3c46c4dd6f38a1a1c611be0521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0