Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000411666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000411666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000411666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000411666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000415417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000415417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000415417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000415417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000453236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
ENST00000453236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
NR_108045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
NR_108045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
NR_108045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009043.hg.1 |
NR_108045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000411666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000411666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000411666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000411666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000415417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000415417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000415417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000415417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000445161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000453236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
ENST00000453236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
NR_108045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
NR_108045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
NR_108045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009043.hg.1 |
NR_108045.1 |
Refseq |