Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00414476_m1
          See other DEFB104A GE Assays ›
          Gene Symbol:
          DEFB104A DEFB104B
          Gene Name:
          defensin beta 104A
          defensin beta 104B
          Gene Aliases:
          BD-4, DEFB-4, DEFB104, DEFB4, hBD-4
          BD-4, DEFB-4, hBD-4
          Chromosome Location:
          Chr.8: 7836471 - 7841242 on Build GRCh38
          Chromosome Location:
          Chr.8: 7470308 - 7475082 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00414476_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DEFB104A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080389.2 NP_525128.2 1 - 2 71 112
          GenBank mRNA AJ314834.1 - 1 - 2 71 112
          BC100848.1 - 1 - 2 71 40002160 112
          BC100849.1 - 1 - 2 71 40002161 112
          BC100850.1 - 1 - 2 71 40002162 112
          BC100851.1 - 1 - 2 71 40002163 112
          Gene Symbol: DEFB104B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001040702.1 NP_001035792.1 1 - 2 71 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800009466.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000922.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AJ314834.1,BC100848.1,BC100849.1,BC100850.1,BC100851.1,NM_001040702.1,NM_080389.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DEFB104A
          Click here to find siRNAs for Gene DEFB104B

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s)

          defense response to bacterium
          innate immune response

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 942610 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 599014 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 761578 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 309414 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 127275 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 243783 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 122894 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 317380 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 361282 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 280871 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 56008 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 264571 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P9018 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P9018 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2996 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2997 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17241 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19041 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P9018 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P9018 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P9018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26217384 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05941584 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16062157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26219086 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05941592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05941591 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26217384 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05941584 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16062157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05941592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26219086 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05941591 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P9018 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05941591 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05941592 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05941584 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P9018 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 93261 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 66144 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8149193 - -
          Affymetrix Human Gene 1.0 ST Array 8144459 - -
          Affymetrix Human Gene 1.0 ST Array 8144459 - -
          Affymetrix Human Gene 1.0 ST Array 8144458 - -
          Affymetrix Human Gene 1.0 ST Array 8144458 - -
          Affymetrix Human Gene 1.0 ST Array 8149194 - -
          Affymetrix Human Gene 1.0 ST Array 8149194 - -
          Affymetrix Human Gene 1.0 ST Array 8149193 - -
          Affymetrix Human Gene 1.1 ST Array 8144457 - -
          Affymetrix Human Gene 1.1 ST Array 8144457 - -
          Affymetrix Human Gene 1.1 ST Array 8149192 - -
          Affymetrix Human Gene 1.1 ST Array 8149192 - -
          Affymetrix Human Transcriptome Array 2.0 PSR08012013.hg.1 TC08000922.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08012013.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08012013.hg.1 NM_001040702.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08012013.hg.1 NM_080389.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006621.hg.1 ENST00000314265.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006621.hg.1 ENST00000314265.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006621.hg.1 ENST00000314265.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08012014.hg.1 TC08000922.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08012014.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08012014.hg.1 NM_001040702.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08012014.hg.1 NM_080389.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009466.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009466.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009466.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009466.hg.1 NM_080389.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009466.hg.1 NM_080389.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009466.hg.1 NM_080389.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08012012.hg.1 TC08000922.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08012012.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08012012.hg.1 NM_001040702.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08012012.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800164756.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164756.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800164756.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164756.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800164756.hg.1 TC0800009466.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800006621.hg.1 ENST00000314265.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006621.hg.1 ENST00000314265.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006621.hg.1 ENST00000314265.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164755.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164755.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800164755.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164755.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800164755.hg.1 TC0800009466.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800164753.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164753.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800164753.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164753.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800164753.hg.1 TC0800009466.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800009466.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009466.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009466.hg.1 ENST00000316169.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009466.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay TC0800009466.hg.1 NM_080389.2 Refseq
          Affymetrix Human Clariom D Assay TC0800009466.hg.1 NM_080389.2 Refseq
          Human Genome U133 Plus 2.0 Array 1553521_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553521_at - -
          Human Genome U133 Plus 2.0 Array 1553521_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553521_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553521_at7 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036315 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036315 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036315 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036315 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 242068 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1725401 AJ314834.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2177234 NM_001040702.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2177234 AJ314834.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2177234 BC100848.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2177234 BC100849.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2177234 NM_080389.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1725401 AJ314834.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2177234 NM_001040702.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2177234 AJ314834.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2177234 BC100848.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2177234 BC100849.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2177234 NM_080389.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1725401 AJ314834.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2177234 NM_001040702.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2177234 AJ314834.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2177234 BC100848.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2177234 BC100849.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2177234 NM_080389.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2177234 NM_001040702.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2177234 AJ314834.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2177234 BC100848.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2177234 BC100849.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2177234 NM_080389.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_17939347-S NM_001040702.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_17939347-S AJ314834.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_17939347-S BC100848.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_17939347-S BC100849.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_17939347-S NM_080389.2 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_2174-S - -
          Illumina human-6 v1.0 expression beadchip 0003170593 - -
          Illumina humanRef-8 v1.0 expression beadchip 0003170593 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1725401 AJ314834.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1725401 AJ314834.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1240494 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_2174 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2177234 NM_001040702.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2177234 AJ314834.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2177234 BC100848.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2177234 BC100849.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2177234 NM_080389.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1725401 AJ314834.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2177234 NM_001040702.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2177234 AJ314834.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2177234 BC100848.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2177234 BC100849.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2177234 NM_080389.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2177234 NM_001040702.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2177234 AJ314834.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2177234 BC100848.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2177234 BC100849.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2177234 NM_080389.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1725401 AJ314834.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1725401 AJ314834.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2177234 NM_001040702.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2177234 AJ314834.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2177234 BC100848.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2177234 BC100849.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2177234 NM_080389.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0