Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015702.hg.1 |
ENST00000271277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015702.hg.1 |
ENST00000496194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015702.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015702.hg.1 |
TC01000987.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000502356.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000502356.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000502356.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000607039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000607039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015700.hg.1 |
ENST00000271277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015700.hg.1 |
ENST00000496194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015700.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015700.hg.1 |
TC01000987.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015693.hg.1 |
ENST00000271277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015693.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015693.hg.1 |
TC01000987.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015701.hg.1 |
ENST00000271277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015701.hg.1 |
ENST00000496194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015701.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015701.hg.1 |
TC01000987.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000502356.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000502356.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000502356.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000607039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000607039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009441.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167936.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167936.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167936.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167936.hg.1 |
TC0100009441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100167936.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167936.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167936.hg.1 |
ENST00000441739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167930.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167930.hg.1 |
NM_018704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167930.hg.1 |
TC0100009441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100167930.hg.1 |
ENST00000271277.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167930.hg.1 |
NM_018704.2 |
Refseq |