Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00236874_m1
          See other LTA GE Assays ›
          Gene Symbol:
          LTA
          Gene Name:
          lymphotoxin alpha
          Gene Aliases:
          LT, TNFB, TNFSF1, TNLG1E
          Chromosome Location:
          Chr.6: 31560550 - 31574324 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00236874_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LTA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000595.3 NP_000586.2 3 - 4 372 132
          NM_001159740.2 NP_001153212.1 3 - 4 423 132
          XM_011514615.2 XP_011512917.1 6 - 7 1307 132
          XM_011514616.2 XP_011512918.1 4 - 5 1042 132
          XM_011514617.2 XP_011512919.1 6 - 7 909 132
          XM_011514618.1 XP_011512920.1 6 - 7 855 132
          GenBank mRNA A06316.1 - 3 - 4 249 132
          BC034729.1 - 2 - 3 216 4341621 132
          BG685356.1 - 2 - 3 459 132
          BP338937.1 - 3 - 4 423 132
          CR542253.1 - 2 - 3 210 132
          CR542273.1 - 2 - 3 210 132
          D00102.1 - 3 - 4 272 132
          D12614.1 - 3 - 4 350 132
          DQ123824.1 - 1 - 2 66 132
          DQ123825.2 - 1 - 2 66 132
          DQ123826.1 - 1 - 2 66 132
          X01393.1 - 3 - 4 289 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007595.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000370.hg.1, TC6_cox_hap2000066.hg.1, TC6_dbb_hap3000057.hg.1, TC6_mann_hap4000058.hg.1, TC6_mcf_hap5000052.hg.1, TC6_qbl_hap6000057.hg.1, TC6_ssto_hap7000053.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human Immune Response Human NFkB Pathway Human TNFR2 Pathway Human Cytokine Network Human IL-9 PATHWAY Human TNF Superfamily Pathway HUMAN PATHOGENESIS OF RHEUMATOID ARTHRITIS Inventoried MicroArray Quality Control (MAQC) Human_Immune Human_IL-9_Pathway Human_Nfkb Human_TNFR2_Pathway Human_Cytokine_Network Human_TNF_Superfamily_Pathway Human_Pathogenesis_of_Rheumatoid_Arthritis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LTA

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of chronic inflammatory response to antigenic stimulus
          positive regulation of humoral immune response mediated by circulating immunoglobulin
          apoptotic process
          humoral immune response
          signal transduction
          cell-cell signaling
          response to nutrient
          response to lipopolysaccharide
          positive regulation of interferon-gamma production
          tumor necrosis factor-mediated signaling pathway
          response to drug
          positive regulation of apoptotic process
          negative regulation of growth of symbiont in host
          negative regulation of fibroblast proliferation
          lymph node development
          defense response to Gram-positive bacterium
          positive regulation of glial cell proliferation
          receptor binding
          cytokine activity
          tumor necrosis factor receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000418386.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000454783.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000471842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 ENST00000489638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_000595.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007595.hg.1 NM_001159740.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0