Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00233632_m1
          See other FMR1 GE Assays ›
          Gene Symbol:
          FMR1
          Gene Name:
          fragile X mental retardation 1
          Gene Aliases:
          FMRP, FRAXA, POF, POF1
          Chromosome Location:
          Chr.X: 147911951 - 147951127 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00233632_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FMR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001185075.1 NP_001172004.1 12 - 13 1416 81
          NM_002024.5 NP_002015.1 12 - 13 1416 81
          NR_033699.1 12 - 13 1416 81
          GenBank mRNA KJ534837.1 - 12 - 13 1187 81
          KM091457.1 - 13 - 14 1295 81
          KM091459.1 - 13 - 14 1295 81
          S65791.1 - 12 - 13 1406 81
          X69962.1 - 12 - 13 1406 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00008668.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000705.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Immune Tolerance Network Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FMR1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          positive regulation of receptor internalization
          mRNA processing
          cellular response to DNA damage stimulus
          glutamate receptor signaling pathway
          RNA splicing
          negative regulation of translation
          gene silencing by RNA
          positive regulation of histone phosphorylation
          cellular response to UV
          regulation of mRNA stability
          modulation by host of viral RNA genome replication
          positive regulation of translation
          negative regulation of translational initiation
          regulation of neurotransmitter secretion
          mRNA transport
          regulation of filopodium assembly
          positive regulation of filopodium assembly
          regulation of gene silencing by miRNA
          regulation of dendritic spine development
          positive regulation of dendritic spine development
          cellular response to hydroxyurea
          cellular response to virus
          regulation of neuronal action potential
          negative regulation of long-term synaptic depression
          positive regulation of intracellular transport of viral material
          negative regulation of voltage-gated calcium channel activity
          positive regulation of proteasomal protein catabolic process
          positive regulation of mRNA binding
          negative regulation of synaptic vesicle exocytosis
          positive regulation of gene silencing by miRNA
          negative regulation of cytoplasmic translation
          positive regulation of response to DNA damage stimulus
          G-quadruplex RNA binding
          chromatin binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          protein binding
          microtubule binding
          poly(U) RNA binding
          translation repressor activity
          translation initiation factor binding
          RNA strand annealing activity
          poly(G) binding
          methylated histone binding
          siRNA binding
          miRNA binding
          RNA stem-loop binding
          identical protein binding
          protein homodimerization activity
          ribosome binding
          transmembrane transporter binding
          protein heterodimerization activity
          mRNA 5'-UTR binding
          dynein complex binding
          sequence-specific mRNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 ENST00000218200.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 ENST00000370470.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 ENST00000370471.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 ENST00000370475.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 ENST00000370477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 ENST00000439526.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 ENST00000440235.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 ENST00000492846.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 NM_001185075.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 NM_001185076.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 NM_001185081.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 NM_001185082.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 NM_002024.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 NR_033699.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 NR_033700.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X009025.hg.1 TC0X000705.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000218200.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000370470.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000370471.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000370475.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000370477.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000439526.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000440235.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000492846.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000611273.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000616382.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000620828.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000621453.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_001185075.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_001185076.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_001185081.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_001185082.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_002024.5 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NR_033699.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NR_033700.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 TC0X00008668.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000218200.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000370470.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000370471.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000370475.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000370477.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000439526.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000440235.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000492846.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000611273.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000616382.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000620828.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 ENST00000621453.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_001185075.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_001185076.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_001185081.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_001185082.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NM_002024.5 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NR_033699.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00158702.hg.1 NR_033700.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0