Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00199656_m1
          See other NUDT21 GE Assays ›
          Gene Symbol:
          NUDT21
          Gene Name:
          nudix hydrolase 21
          Gene Aliases:
          CFIM25, CPSF5
          Chromosome Location:
          Chr.16: 56429136 - 56451349 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00199656_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NUDT21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007006.2 NP_008937.1 1 - 2 260 126
          GenBank mRNA AJ001810.1 - 1 - 2 113 126
          AK312721.1 - 1 - 2 263 126
          AL535307.3 - 1 - 2 260 126
          BC001403.2 - 1 - 2 221 3139379 126
          BX537360.1 - 1 - 2 264 126
          BX647665.1 - 1 - 2 246 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600010407.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16001133.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NUDT21

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          termination of RNA polymerase II transcription
          mRNA polyadenylation
          mRNA processing
          mRNA 3'-end processing
          protein tetramerization
          RNA binding
          mRNA binding
          protein binding
          hydrolase activity
          mRNA 3'-UTR AU-rich region binding
          protein homodimerization activity
          histone deacetylase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16015787.hg.1 ENST00000300291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015787.hg.1 ENST00000535563.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015787.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015787.hg.1 TC16001133.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16015786.hg.1 ENST00000300291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015786.hg.1 ENST00000535563.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015786.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015786.hg.1 TC16001133.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000563362.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000563362.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000563860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000563860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000563860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000568822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000568822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000568822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000612596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000612596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 ENST00000612596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012440.hg.1 ENST00000447151.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012440.hg.1 ENST00000447151.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012440.hg.1 ENST00000447151.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 ENST00000568822.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 TC1600010407.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 ENST00000568822.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179893.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179895.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179895.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179895.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179895.hg.1 TC1600010407.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600179895.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179895.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179895.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179897.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179897.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179897.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179897.hg.1 TC1600010407.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600179897.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179897.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179897.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179896.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179896.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179896.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179896.hg.1 TC1600010407.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600179896.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179896.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179896.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1100012440.hg.1 ENST00000447151.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012440.hg.1 ENST00000447151.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012440.hg.1 ENST00000447151.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000300291.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000563362.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000563362.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000563860.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000563860.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000563860.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000566340.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000567110.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000568822.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000568822.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000568822.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000612596.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000612596.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 ENST00000612596.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010407.hg.1 NM_007006.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 100313 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0