Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00171267_m1
          See other EPO GE Assays ›
          Gene Symbol:
          EPO
          Gene Name:
          erythropoietin
          Gene Aliases:
          EP, MVCD2
          Chromosome Location:
          Chr.7: 100720800 - 100723700 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00171267_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EPO
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000799.2 NP_000790.2 1 - 2 199 134
          GenBank mRNA X02157.1 - 1 - 2 199 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700008563.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000632.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_000799.2, X02157.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Human Hypoxia Panel Human Erythropoietin Pathway HUMAN JAK-STAT PATHWAY Inventoried MicroArray Quality Control (MAQC) Human_Hypoxia Human_Erythropoietin_Pathway Human_JAK-STAT_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene EPO

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          apoptotic process
          acute-phase response
          signal transduction
          embryo implantation
          aging
          blood circulation
          positive regulation of cell population proliferation
          response to salt stress
          negative regulation of calcium ion transport into cytosol
          positive regulation of neuron projection development
          peptidyl-serine phosphorylation
          erythrocyte differentiation
          activation of protein kinase activity
          response to lipopolysaccharide
          response to vitamin A
          response to testosterone
          positive regulation of activated T cell proliferation
          positive regulation of tyrosine phosphorylation of STAT protein
          hemoglobin biosynthetic process
          erythrocyte maturation
          response to estrogen
          positive regulation of DNA replication
          positive regulation of transcription, DNA-templated
          positive regulation of Ras protein signal transduction
          response to axon injury
          response to electrical stimulus
          response to hyperoxia
          regulation of transcription from RNA polymerase II promoter in response to hypoxia
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-1
          cellular hyperosmotic response
          negative regulation of neuron death
          negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
          negative regulation of erythrocyte apoptotic process
          negative regulation of cation channel activity
          erythropoietin receptor binding
          hormone activity
          protein binding
          protein kinase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07009695.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07009695.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07009695.hg.1 TC07000632.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07009696.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07009696.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07009696.hg.1 TC07000632.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07009694.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07009694.hg.1 NM_000799.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07009694.hg.1 TC07000632.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700160373.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700160373.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700160373.hg.1 TC0700008563.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700160373.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700160373.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay TC0700008563.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700160374.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700160374.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700160374.hg.1 TC0700008563.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700160374.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700160374.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700160376.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700160376.hg.1 NM_000799.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700160376.hg.1 TC0700008563.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700160376.hg.1 ENST00000252723.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700160376.hg.1 NM_000799.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5db945b66b-542qb:80/100.66.134.79:80.
          git-commit: 84360441031e17f3391aad9879d55c303cd12901
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0